missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CGA (Human) Recombinant Protein
Human CGA full-length ORF ( AAH10957, 1 a.a. - 116 a.a.) recombinant protein with GST-tag at N-terminal.
POA
Specifications
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
---|---|
Gene ID (Entrez) | 1081 |
Description
- Sequence: MDYYRKYAAIFLVTLSVFLHVLHSAPDVQDCPECTLQENPLFSQPGAPILQCMGCCFSRAYPTPLRSKKTMLVQKNVTSESTCCVAKSYNRVTVMGGFKVENHTACHCSTCYYHKS
Specifications
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
1081 |