Learn More
Microspec™ Centrifuge Tube with Conical Bottom
Polypropylene centrifuge tube available in non-sterile or EtO sterile.
Brand:  Microspec™ AF2
Additional Details : Weight : 3.10000kg
 View more versions of this product
Product Code. 16645472
  • 121.00 € / Pack of 500
Estimated Shipment: 05-06-2024
to see stock.
Add to basket
Description
Description
  • Conical bottom
  • Flat cap
  • No skirt
  • Polypropylene
Specifications
Specifications
Polypropylene
50 mL
50/Inner Bag
Not Autoclavable
Screw Cap
3000 x g
Blue
500
Sterile
Conical