Learn More
Microspec™ Centrifuge Tube with Conical Bottom
Polypropylene centrifuge tube available in non-sterile or EtO sterile.
Brand: Microspec™ AF2
Additional Details : Weight : 3.10000kg
Description
- Conical bottom
- Flat cap
- No skirt
- Polypropylene
Specifications
Polypropylene | |
50 mL | |
50/Inner Bag | |
Not Autoclavable | |
Screw Cap | |
3000 x g |
Blue | |
500 | |
Sterile | |
Conical | |
Yes |