missing translation for 'onlineSavingsMsg'
Learn More

Manta Diagnostics™ Cellulose Nitrate Filters with Grid

200 filters per pack, individually wrapped

Brand:  Manta Diagnostics™ AL2

Additional Details : Weight : 0.15000kg

 View more versions of this product

Product Code. 16645492

  • 93.64 € / Pack of 200

Estimated Shipment: 11-01-2022
Add to basket

Description

Description

  • Cellulose Nitrate
  • 47 mm Diameter
  • 200 per pack
  • Individually wrapped
  • Sterile
Specifications

Specifications

Cellulose Nitrate
Sterile
47 mm
Yes
1
0.22 μm
Blue
Not Autoclavable
200
Videos
SDS
Documents
Certificates
Promotions

Promotions