missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One CELLCOAT™ Collagen Type I Coated Cell Culture Flask
Ideal for cell cultivation in serum-free or serum-reduced medium
125.00 € - 1400.00 €
Specifications
Closure Color | Red |
---|---|
Closure Material | HDPE |
Material | Polystyrene |
Neck Style | Canted |
Surface Coating | Collagen Type I Protein |
Product Code | Brand | Culture Area | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Culture Area | Price | Quantity & Availability | |||||
11709492
|
Greiner Bio-One
661950 |
175 cm2 |
1400.00 €
Pack of 40 |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
13092263
|
Greiner Bio-One
658950 |
75 cm2 |
959.00 €
Pack of 50 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
10174431
|
Greiner Bio-One
661950 |
175 cm2 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Promotion of cell adhesion, proliferation and growth of endothelial cell, hepatocytes, muscle cells, pheochromocytoma cells (PC12) and other cell types
- Source: Rat tail Collagen Type I
- Ready-to-use
- Supplied with red filter screw cap.
Specifications
Red | |
Polystyrene | |
Collagen Type I Protein | |
Filter Screw Cap |
HDPE | |
Canted | |
Yes |