Learn More
CD33 Monoclonal Antibody (P67.6), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand:  Affymetrix eBioscience 25-0337-41
116.00 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16646314
  • 174.00 € / 25 tests
Estimated Shipment: 15-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description
Description
CD33 Monoclonal antibody specifically detects CD33 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD33
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P20138
CD33
Affinity chromatography
RUO
945
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
P67.6
PE-Cyanine7
CD33
CD33; CD33 antigen; CD33 antigen (gp67); CD33 molecule; CD33 molecule transcript; FLJ00391; gp67; Myeloid cell surface antigen CD33; p67; sialic acid binding Ig-like lectin 3; sialic acid-binding Ig-like lectin 3; SIGLEC3; Siglec-3
Mouse
25 Tests
Primary
Human
Antibody
IgG1 κ