missing translation for 'onlineSavingsMsg'
Learn More

CD195 (CCR5) Mouse anti-Human, Alexa Fluor 488, Clone: NP-6G4, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-1956-42

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16646404

  • 287.00 € / 100 tests

Estimated Shipment: 09-12-2021
Add to basket

Description

Description

Description: The NP-6G4 monoclonal antibody reacts with human CD195 (CCR5), an approximately 45 kDa member of the G protein-coupled receptor (GPCR) superfamily that binds chemokines including MIP-1a, MIP-1b and RANTES. Human CCR5 is expressed on resting T cells with a memory/effector phenotype, monocytes, macrophages and immature dendritic cells. CCR5 functions as a receptor to direct the chemotaxis of lymphocytes, and is the primary co-receptor for macrophage-tropic Human Immunodeficiency Virus Type 1 (HIV-1), which binds CCR5 through gp120. Applications Reported: This NP-6G4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This NP-6G4 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. For optimal performance, it is recommended that staining with NP-6G4 be done at 37°C for 1 hour in Flow Cytometry Staining Buffer (Product # 00-4222-26). This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser

Human immunodeficiency virus (HIV) and related virus require coreceptors, in addition to CD4, to infect target cells. Some G protein-coupled receptors including CCR5, CXCR4, CCR3, CCR2b and CCR8 in the chemokine receptor family, and four new human molecules GPR15, STRL33, GPR1 and V28 were recently identified as HIV coreceptors. Among them, CCR5 (CC-CKR-5) is a principal coreceptor for macrophage- and dual-tropic HIV-1 strains fusion and entry of human white blood cells. CCR5 is required for the infection by HIV-1, HIV-2, and SIV. The beta-chemokines RANTES, MIP-alpha and MIP-beta are the ligands for CCR5 and prevent infection by M-tropic HIV-1. CCR5 associates with the surface CD4-gp120 of HIV complex and leads to membrane fusion and virus entry of target cells. The amino-terminal domain and the extracellular loops of CCR5 serve as HIV biding sites. CCR5 messenger RNA is expressed in lymphoid organs and monocytes.
Specifications

Specifications

CD195 (CCR5)
NP-6G4
Alexa Fluor 488
CCR5
Liquid
CCR5
IgG1, kappa
100 Tests
4°C, store in dark, DO NOT FREEZE!
Primary
1234
Flow Cytometry
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P51681
AM4-7; C-C chemokine receptor type 5; C-C CKR-5; C-C motif chemokine receptor 5 (gene/pseudogene); C-C motif chemokine receptor 5 A159A; CCCKR5; CC-CKR-5; Ccr5; CCR-5; CD195; CD195 antigen; chemokine (C-C motif) receptor 5; chemokine (C-C) receptor 5; chemokine receptor CCR5; CHEMR13; CKR5; CKR-5; CMKBR5; HIV-1 fusion coreceptor; IDDM22; MIP-1 alpha receptor
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Videos
SDS
Documents
Certificates
Promotions

Promotions