missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 Monoclonal Antibody (TuK4), Alexa Fluor™ 647, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A51013
Additional Details : Weight : 0.00460kg
Description
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry.Specifications
CD14 | |
Monoclonal | |
0.04 mg/mL | |
PBS with 1% BSA and 0.0325% sodium azide; pH 7.4 | |
P08571 | |
Cd14 | |
Human CD14 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry | |
TuK4 | |
Alexa Fluor 647 | |
Cd14 | |
CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
Mouse | |
Affinity chromatography | |
RUO | |
929 | |
4° C, store in dark | |
Liquid |