Learn More
Thermo Scientific™ CaptureSelect™ C-tagXL Pre-packed Column
Thermo Scientific™ CaptureSelect™ C-tagXL Affinity Matrix is an improved version of CaptureSelect C-tag Affinity Matrix with a significantly increased binding capacity, offered here in pre-packed format.
1515.00 €
Specifications
Content And Storage | Store in refrigerator (2°C to 8°C). |
---|---|
Shipping Condition | Room Temperature |
Form | Liquid Suspension |
Product Line | CaptureSelect™ |
Type | Pre-packed Column |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18476458
|
Thermo Scientific™
494307205 |
1 x 5 mL Column |
1515.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18416528
|
Thermo Scientific™
494307201 |
5 x 1 mL Columns |
1515.00 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CaptureSelect C-tagXL Affinity Matrix is an improved version of CaptureSelect C-tag Affinity Matrix with a significantly increased binding capacity, offered here in pre-packed format.
Like C-tag Affinity Matrix, C-tagXL Affinity Matrix combines a unique selectivity for a small 4-amino acid peptide tag (E-P-E-A, glutamic acid-proline-glutamic acid-alanine) with the benefits of a robust and high quality affinity matrix. CaptureSelect C-tagXL Affinity Matrix purifies C-terminal tagged proteins with high affinity and selectivity, even in the presence of urea and guanidine HCl, from complex mixtures like cell culture harvests and periplasmatic fractions in a one-step process. Mild elution conditions at neutral pH can be applied using magnesium chloride or propylene glycol, which ensures high activity recoveries of pH-sensitive target proteins. The affinity resin recognizes the E-P-E-A tag sequence when fused either directly to the C-terminus of a protein or through a linker between the C-terminus and the E-P-E-A tag.
The affinity matrix is offered in a pre-packed format as a set of 5 x 1 mL columns, or as one 5 mL column, with connectors that are compatible with standard chromatography equipment.
Specifications
Store in refrigerator (2°C to 8°C). | |
Liquid Suspension | |
Pre-packed Column |
Room Temperature | |
CaptureSelect™ | |
Anti-C-tagXL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.