Learn More
Erlab™ Captair Smart Filtered Storage Cabinets
New Smart Technology uses simple light to show that the hood is operating safely so the user can focus their attention on what is most important: their work. When the light is still the hood is operating safely. To alert the user, the light will pulse associated to an acoustic alarm.
POA
Specifications
Product Type Filtered Storage Cabinet
View More Specs
Description
Description
  • New Smart Technology: designed with a simple and innovative way of communication
  • Smart Technology uses simple light to show that the hood is operating safely so you can focus your attention on what is most important: your work. When the light is still the hood is operating safely. To alert you, the light will pulse associated to an acoustic alarm
  • A simple light pattern will indicate if : air flow is compromised, fan failure, filter breakthrough, doors are left open
  • The new eGuard App gives you remote control to monitor the hood, change the settings, and delivers safety alerts immediately to your mobile, tablet or PC device
  • Configurable filtering system above the enclosure can be adapted to filter gases, solvents, powders and particulates keeping the user and the lab protected by safely and efficiently recirculating air within the room and releasing purified air back into the room free of any toxic hazardous vapours or odours
  • Meets AFNOR NFX 15 211/ANSI Z9.5-2012 filtration efficiency standard (classes 1 and 2)
  • New sensors are added for detection of solvents or acids or formaldehyde.
  • New sensors are added if doors are left open.
  • ESP Program: Usage certification delivered with each unit after approval of handlings or process by the manufacturer
  • Very low energy consumption, only 60W
  • Smaller sizes are available for use on or under counters or equipment.
  • Floor-standing models can be configured with adjustable spill-proof shelves, pull-out vertical shelves, and large volume drawers
Specifications
Specifications
Filtered Storage Cabinet