missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, Free-flowing, ACS reagent, anhydrous, ≥96%, Honeywell Fluka™

Catalog Number p-7111991
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
10043-52-4
CaCl2
110.98
MFCD00010903
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
5284359
CHEBI:3312
calcium;dichloride
[Cl-].[Cl-].[Ca++]
This item is not returnable. View return policy
10043-52-4
CaCl2
110.98
MFCD00010903
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
5284359
CHEBI:3312
calcium;dichloride
[Cl-].[Cl-].[Ca++]

Free-flowing, ACS reagent, anhydrous,≥96%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Boiling Point 1,900°C
Melting Point 772°C (literature)
CAS Max % 100.0000%
Physical Form Anhydrous
Quantity 100 g
Linear Formula CaCl2
Formula Weight 110.98
Percent Purity >96%
Grade ACS Reagent
Packaging Poly Bottle
Density 2.150 g/cm3
Chemical Name or Material Calcium chloride, Free-flowing, ACS reagent, anhydrous, ≥96%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'