missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Carbonate, Powder, Low in Alkalies, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Specifications
CAS | 471-34-1 |
Physical Form | Powder |
Quantity | 500 g |
Molecular Formula | CCaO3 |
MDL Number | MFCD00010906 |
Synonym | calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama |
InChI Key | VTYYLEPIZMXCLO-UHFFFAOYSA-L |
SMILES | [Ca++].[O-]C([O-])=O |
IUPAC Name | calcium;carbonate |
Molecular Weight (g/mol) | 100.09 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'