missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium for AAS 1g/L HCl matrix, Fisher chemical™
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Bottle
Specifications
Concentration | 1g/L ±0.002g/L |
Packaging | Bottle |
Quantity | 500 mL |
Type | Flame Single-element Standard Solution |
UN Number | 3082 |
Solvent or Matrix | 2% HCl |
Physical Form | Liquid |
Chemical Name or Material | Cadmium |
Safety and Handling
Product Identifier
- Single-element Standard Solution for AAS. Cadmium (Cd) 1000mg/l in HCl 2% (B 010 )
- Danger
- Acute toxicity Category 4
- Germ cell mutagenicity Category 1B
- Carcinogenicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'