missing translation for 'onlineSavingsMsg'
Learn More

BX471 hydrochloride, MedChemExpress

Product Code. p-200099718
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16474978

Brand: Medchem Express HY12080A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BX471 hydrochloride (ZK-811752 hydrochloride) is a potent, selective non-peptide CCR1 antagonist with Ki of 1 nM for human CCR1, and exhibits 250-fold selectivity for CCR1 over CCR2, CCR5 and CXCR4.

BX471 hydrochloride (ZK-811752 hydrochloride) is a potent, selective non-peptide CCR1 antagonist with Ki of 1 nM for human CCR1, and exhibits 250-fold selectivity for CCR1 over CCR2, CCR5 and CXCR4.

Specifications

Chemical Name or Material BX471 hydrochloride
CAS 288262-96-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H25Cl2FN4O3
Quantity 10 mg
Synonym ZK-811752 hydrochloride
Solubility Information DMSO : 150 mg/mL (318.23 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES O=C(N)NC1=CC(Cl)=CC=C1OCC(N2[C@H](C)CN(CC3=CC=C(F)C=C3)CC2)=O.[H]Cl
Molecular Weight (g/mol) 471.35
Formula Weight 471.35
Percent Purity 99.51%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.