missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 7.413, Fisher Chemical™
Click to view available options
Quantity:
500 mL
1000 mL
Specifications
pH | 7.413 |
Physical Form | Liquid |
Quantity | 500 mL |
Solution Type | Buffer |
Formulation | Phosphate Potassium, Dihydrogen/Phosphate di-Sodium Hydrogen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'