Learn More
Buffer Solution, pH 2.00 (Certified), Fisher Chemical™
Brand: Fisher Chemical SB96-20
Additional Details : CAS : 50-00-0 Weight : 20.00000kg
Specifications
Buffer Solution, pH 2.00 | |
7447-40-7,7647-01-0,50-00-0,67-56-1,7732-18-5 | |
Undesignated | |
2.00 | |
20 L | |
MFCD00146206 | |
Traceable to NIST. |
0°C | |
100°C | |
1.0g/cm³ | |
Liquid | |
In Pass Test | |
PolyPac™ | |
Certified |