Learn More
Paragon Scientific™ BS/U/M Miniature Viscometer for Transparent Liquids
For the determination of kinematic viscosity of transparent Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
177.69 €
Specifications
Calibrated | No |
---|---|
ASTM Numbers | D 445, D 446 |
Certifications/Compliance | ASTM D 445, D 446 and ISO 3104, 3105 |
Product Code | Brand | Viscosity Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Viscosity Range | Price | Quantity & Availability | |||||
16030240
|
Paragon Scientific™
MINIU01NC |
0.2 to 1 cSt |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16050240
|
Paragon Scientific™
MINIU02NC |
1 to 5 cSt |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16070240
|
Paragon Scientific™
MINIU03NC |
3 to 15 cSt |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16090240
|
Paragon Scientific™
MINIU04NC |
8 to 40 cSt |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16010250
|
Paragon Scientific™
MINIU05NC |
20 to 100 cSt |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- High Quality glass capillary viscometer
- Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
No | |
D 445, D 446 | |
ASTM D 445, D 446 and ISO 3104, 3105 |
4 week lead-time