missing translation for 'onlineSavingsMsg'
Learn More

Boc-MLF TFA, MedChemExpress

Product Code. 16420227
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16420227

Brand: Medchem Express HY103473A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Boc-MLF (TFA) is a peptide, used as a specific formyl peptide receptor (FPR) antagonist, also inhibits the signaling through formyl peptide receptor like 1 (FPRL1) at higher concentrations.

Boc-MLF (TFA) is a peptide, used as a specific formyl peptide receptor (FPR) antagonist, also inhibits the signaling through formyl peptide receptor like 1 (FPRL1) at higher concentrations.

Specifications

Chemical Name or Material Boc-MLF TFA
Physical Form Solid
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C27H40F3N3O8S
For Use With (Application) COVID-19-immunoregulation
Synonym Boc-Met-Leu-Phe-OH (TFA)
Solubility Information DMSO : 150 mg/mL (240.51 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(O)[C@H](CC1=CC=CC=C1)NC([C@H](CC(C)C)NC([C@H](CCSC)NC(OC(C)(C)C)=O)=O)=O.OC(C(F)(F)F)=O
Molecular Weight (g/mol) 623.68
Formula Weight 623.68
Percent Purity 99.7%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.