missing translation for 'onlineSavingsMsg'
Learn More

Bis(di-tert-butyl-phenylphosphine)palladium(0), 98%, Thermo Scientific Chemicals

Catalog Number p-7059983
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
52359-17-8
C28H46P2Pd
551.04
MFCD15071400
KJTQUBRAVOMHKY-UHFFFAOYSA-N
palladium, bis bis 1,1-dimethylethyl phenylphosphine, di-tert-butyl phenyl phosphane-palladium 2/1, bis di-tert-butyl phenyl phosphane palladium
71365671
ditert-butyl(phenyl)phosphane;palladium
[Pd].CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C
This item is not returnable. View return policy
52359-17-8
C28H46P2Pd
551.04
MFCD15071400
KJTQUBRAVOMHKY-UHFFFAOYSA-N
palladium, bis bis 1,1-dimethylethyl phenylphosphine, di-tert-butyl phenyl phosphane-palladium 2/1, bis di-tert-butyl phenyl phosphane palladium
71365671
ditert-butyl(phenyl)phosphane;palladium
[Pd].CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 551.04
Concentration or Composition (by Analyte or Components) Pd 19.3%
Physical Form Crystalline
Chemical Name or Material Bis (di-tert-butyl-phenylphosphine)palladium (0)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'