Learn More
Ansell BioClean™ Durableu Disposable Overshoes BDBO
Catalog Number p-7162364
Click to view available options
Size:
Standard
X-Large
Ansell™ BioClean™ Durableu Disposable Overshoes BDBO offer superb protection and durability.
  • Resists tears, rips, and abrasions
  • Low levels of particle shedding
  • Resistant to a wide range of liquid chemicals
  • High tensile strength
  • Slip-resistant
  • Non-woven spunbonded polypropylene (38 gsm)/Cast polyethylene laminate (64 gsm)
Size Standard
Material Polypropylene
Latex Free No
Color Blue
Disposable Single-use
Sterility Non-sterile
Certifications/Compliance Manufactured in a facility holding ISO 9001:2015
Packaging Quantity 50 Pieces per Sealed Inner PE Bag, 1 Inner Bag per Sealed Outer PE Bag, 8 Outer Bags per Lined Carton
Packaging Type 400/Cs.
Product Type Controlled Environments Shoe Covers
Five (5) years from date of manufacture. Store in a dry, cool place (≤40°C) away from direct sunlight and fluorescent light.