missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals beta-Casomorphin, human, 95+%

Catalog Number p-7058218
Click to view available options
Quantity:
5mg
25mg
This item is not returnable. View return policy
102029-74-3
This item is not returnable. View return policy
102029-74-3

An opioid peptide

An opioid peptide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Purity ≥95%
Color White
Content And Storage Store at -20°C
Format Powder
Molecular Weight (g/mol) 864.01
Product Type beta-Casomorphin
Quantity 5mg

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'