Learn More
SHOWA™ 890 Unlined Viton™ Chemical Resistant Butyl Gloves
Click to view available options
Size 10
Description
Extra-thick 14 in. chemical resistant gloves provide resistance to highly corrosive acids. Butyl rubber coating is designed for handling aliphatic and aromatic hydrocarbons, such as Benzene, Toulene, and Xylene.
- Forged with fluoroelastomer rubber
- Butyl rubber coating
- Hydrocarbon-resistant
- Impermeable
- Forearm protection
- Resistant to highly corrosive acids
- Smooth surface grip allows for increased sensitivity and tactility