missing translation for 'onlineSavingsMsg'
Learn More

Benzyldimethylhexadecylammonium chloride hydrate, 97%

Product Code. p-66010 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
Unit Size:
500g
5g
This item is not returnable. View return policy

Product Code. 10556972

Brand: Thermo Scientific Acros 402070050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122-18-9 | C25H46ClN | 396.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-18-9
Molecular Formula C25H46ClN
Molecular Weight (g/mol) 396.10
MDL Number MFCD00149967
InChI Key SXPWTBGAZSPLHA-UHFFFAOYSA-M
Synonym cetalkonium chloride, benzyldimethylhexadecylammonium chloride, banicol, bonjela, cetyl zephiran, cetyldimethylbenzylammonium chloride, bicetonium, acinol, benzylhexadecyldimethylammonium chloride, baktonium
PubChem CID 31202
IUPAC Name benzyl-hexadecyl-dimethylazanium;chloride
SMILES [Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 54.0°C to 58.0°C
Color Cream-White to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (on anhydrous substance) (Argentometry)
Packaging Glass bottle
Quantity 5 g
Beilstein 12, III, 2212
Merck Index 15, 2016
Solubility Information Solubility in water: 8.5g/L. Other solubilities: soluble in alcohol, acetone, esters, ethyla acetat, e, propylene glycol, sorbitol solutions, glycerol,, ether
Formula Weight 396.1
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Benzyldimethylhexadecylammonium chloride hydrate, 97 - 101%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzyldimethylhexadecylammonium chloride, hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.