Learn More
Invitrogen™ BenchPro™ 2100 Piercing Device
The BenchPro™ 2100 Piercing Device is used with the BenchPro™ 2100 Plasmid Purification Card and Reagent Tray Kit (Cat No. MC2001).
Brand:  Invitrogen™ MC3001
Additional Details : Weight : 0.44000kg
Product Code. 11370972
  • 25.49 € / Each
Estimated Shipment: 16-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
For Research Use Only. All usage must comply with product instructions.
Description
Description
The BenchPro™ 2100 Piercing Device is used with the BenchPro™ 2100 Plasmid Purification Card and Reagent Tray Kit (Cat No. MC2001). It is used to pierce the foil seal covering the reagent tray prior to use.
Specifications
Specifications
1 piercing device (for use with MC2001)
BenchPro™
1 Ea.
BenchPro™ 2100 Plasmid Purification System
2100 Piercing Device
Room Temperature