Learn More
Invitrogen™ BenchPro™ 2100 Piercing Device
The BenchPro™ 2100 Piercing Device is used with the BenchPro™ 2100 Plasmid Purification Card and Reagent Tray Kit (Cat No. MC2001).
Brand: Invitrogen™ MC3001
Additional Details : Weight : 0.44000kg
Description
The BenchPro™ 2100 Piercing Device is used with the BenchPro™ 2100 Plasmid Purification Card and Reagent Tray Kit (Cat No. MC2001). It is used to pierce the foil seal covering the reagent tray prior to use.Specifications
1 piercing device (for use with MC2001) | |
BenchPro™ | |
1 Ea. |
BenchPro™ 2100 Plasmid Purification System | |
2100 Piercing Device | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.