missing translation for 'onlineSavingsMsg'
Learn More

Bathophenanthroline, 98+%

Product Code. p-7021782
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11452397

Brand: Thermo Scientific Alfa Aesar A14258.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 1662-01-7 | C24H16N2 | 332.406 g/mol

Bathophenanthroline is widely used for determination of iron in serum and urine by colorimtetry. It acts as buffer layer to improve the efficiency of organic photovoltaic cells. It is very useful as a scavenger reagent, for the removal of traces of iron and copper from reagent solutions, which is used in the trace metal determination. Iron bathophenanthroline complex is involved as a redox mediating agent for imaging surface immobilized deoxyribonucleic acid (DNA) by using Scanning Electrochemical Microscopy (SECM).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bathophenanthroline is widely used for determination of iron in serum and urine by colorimtetry. It acts as buffer layer to improve the efficiency of organic photovoltaic cells. It is very useful as a scavenger reagent, for the removal of traces of iron and copper from reagent solutions, which is used in the trace metal determination. Iron bathophenanthroline complex is involved as a redox mediating agent for imaging surface immobilized deoxyribonucleic acid (DNA) by using Scanning Electrochemical Microscopy (SECM).

Solubility
Soluble in organic solvents. Slightly soluble in acidic aqueous solutions. Insoluble in neutral or alkaline solutions.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1662-01-7
Molecular Formula C24H16N2
Molecular Weight (g/mol) 332.406
MDL Number MFCD00004976
InChI Key DHDHJYNTEFLIHY-UHFFFAOYSA-N
Synonym bathophenanthroline, 1,10-phenanthroline, 4,7-diphenyl, bphen, 1,10-bathophenanthroline, bathophenanthrolin, bathophenanthrolin german, unii-4a2b091f0g, 4,7-diphenyl-1,10-diazaphenanthrene, gnf-pf-4554, 4,7-diphenylpyridino 3,2-h quinoline
PubChem CID 72812
ChEBI CHEBI:77995
IUPAC Name 4,7-diphenyl-1,10-phenanthroline
SMILES C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5

Specifications

Melting Point 218°C to 222°C
Quantity 1 g
Beilstein 261048
Solubility Information Soluble in organic solvents. Slightly soluble in acidic aqueous solutions. Insoluble in neutral or alkaline solutions.
Formula Weight 332.41
Percent Purity ≥98%
Chemical Name or Material Bathophenanthroline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.