Learn More
Bathocuproin, 98%
Descripción
It is used as a reagent for the determination of copper. It acts as an exciton blocking barrier which prohibits excitons diffusion process towards the Al electrode otherwise being quenched. It is the most commonly used buffer layer between acceptor and cathode layer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used as a reagent for the determination of copper. It acts as an exciton blocking barrier which prohibits excitons diffusion process towards the Al electrode otherwise being quenched. It is the most commonly used buffer layer between acceptor and cathode layer.
Solubility
Soluble in methanol (36mg/100ml), ethanol, 2-propanol, bezene, and acetone. Partly miscible in water.
Notes
Store in cool, dry conditions in a sealed container. Store away from oxidizing agents.
Especificaciones
Especificaciones
| Melting Point | 279°C to 285°C |
| Quantity | 1 g |
| Beilstein | 306714 |
| Solubility Information | Soluble in methanol (36mg/100ml),ethanol,2-propanol,bezene,and acetone. Partly miscible in water. |
| Formula Weight | 360.46 |
| Percent Purity | 98% |
| Chemical Name or Material | Bathocuproin |
RUO – Research Use Only
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.