missing translation for 'onlineSavingsMsg'
Learn More

BAPTA tetraethyl ester, 98+%, Thermo Scientific Chemicals

Catalog Number p-7026142
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
73630-07-6
C30H40N2O10
588.654
MFCD00532663
OLXCPQDFHUCXBA-UHFFFAOYSA-N
bapta tetraethyl ester, tetraethyl 2,2',2,2'-ethane-1,2-diylbis oxy bis 2,1-phenylene bis azanetriyl tetraacetate, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid tetraethyl ester, tetraethyl 2,2',2,2'-ethane-1,2-diylbis oxy-2,1-phenylenenitrilo tetraacetate, 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy-n-2-ethoxy-2-oxoethyl anilino acetic acid ethyl ester, ethyl 2-2-2-2-bis 2-ethoxy-2-oxidanylidene-ethyl amino phenoxy ethoxy phenyl-2-ethoxy-2-oxidanylidene-ethyl amino ethanoate, ethyl 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy-n-2-ethoxy-2-oxoethyl anilino acetate, ethyl 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy phenyl 2-ethoxy-2-oxoethyl amino acetate
339960
ethyl 2-[2-[2-[2-[bis(2-ethoxy-2-oxoethyl)amino]phenoxy]ethoxy]-N-(2-ethoxy-2-oxoethyl)anilino]acetate
CCOC(=O)CN(CC(=O)OCC)C1=CC=CC=C1OCCOC2=CC=CC=C2N(CC(=O)OCC)CC(=O)OCC
This item is not returnable. View return policy
73630-07-6
C30H40N2O10
588.654
MFCD00532663
OLXCPQDFHUCXBA-UHFFFAOYSA-N
bapta tetraethyl ester, tetraethyl 2,2',2,2'-ethane-1,2-diylbis oxy bis 2,1-phenylene bis azanetriyl tetraacetate, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid tetraethyl ester, tetraethyl 2,2',2,2'-ethane-1,2-diylbis oxy-2,1-phenylenenitrilo tetraacetate, 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy-n-2-ethoxy-2-oxoethyl anilino acetic acid ethyl ester, ethyl 2-2-2-2-bis 2-ethoxy-2-oxidanylidene-ethyl amino phenoxy ethoxy phenyl-2-ethoxy-2-oxidanylidene-ethyl amino ethanoate, ethyl 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy-n-2-ethoxy-2-oxoethyl anilino acetate, ethyl 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy phenyl 2-ethoxy-2-oxoethyl amino acetate
339960
ethyl 2-[2-[2-[2-[bis(2-ethoxy-2-oxoethyl)amino]phenoxy]ethoxy]-N-(2-ethoxy-2-oxoethyl)anilino]acetate
CCOC(=O)CN(CC(=O)OCC)C1=CC=CC=C1OCCOC2=CC=CC=C2N(CC(=O)OCC)CC(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 98°C to 102°C
Quantity 1 g
Formula Weight 588.66
Percent Purity ≥98%
Chemical Name or Material BAPTA tetraethyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'