missing translation for 'onlineSavingsMsg'
Learn More
Learn More
J.T. Baker Baker-flex™, Cellulose PEI-F, J.T. Baker™
988.00 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
25 Sheets
Specifications
Color | White |
Physical Form | Crystal or Powder |
Solubility Information | Insoluble in water |
Quantity | 25 Sheets |
Odor | Odorless |
Chemical Name or Material | Cellulose PEI-F |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'