Learn More
Abeba™ Autoclavable Clogs 9600
Antistatic and autoclavable at 121°C. High slip resistance.
54.30 € - 58.66 €
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, E SRC |
---|---|
Color | White |
Sole Features | Slip-resistant |
Style | 9600 |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15660456
|
Abeba™
9600/35 |
35 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15670456
|
Abeba™
9600/36 |
36 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15680456
|
Abeba™
9600/37 |
37 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15559080
|
Abeba™
9600/38 |
38 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15690456
|
Abeba™
9600/39 |
39 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15509090
|
Abeba™
9600/40 |
40 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15527580
|
Abeba™
9600/41 |
41 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15557580
|
Abeba™
9600/42 |
42 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15539090
|
Abeba™
9600/43 |
43 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15600466
|
Abeba™
9600/44 |
44 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15559090
|
Abeba™
9600/45 |
45 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15569090
|
Abeba™
9600/46 |
46 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- CE EN ISO 20347:2012 OB A, E SRC
- Autoclavable, sterilisable special material
- White
- Slip resistance outsole sole
- Antistatic
- Heel strap foldable, adjustable
- DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
CE EN ISO 20347:2012 OB A, E SRC | |
White | |
Slip-resistant | |
9600 |