missing translation for 'onlineSavingsMsg'
Learn More

ATP6V1H Recombinant Protein, Abnova

Brand:  Abnova H00051606-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100193

  • 518.16 € / 25µg

Estimated Shipment: 03-01-2022
Add to basket

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH25275
Solution
51606
ATP6V1H (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
CGI-11/MSTP042/NBP1/SFD/SFDalpha/SFDbeta/VMA13
ATP6V1H
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
79.2
8
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATP6V1H
Human
Yes
Videos
SDS
Documents
Certificates
Promotions

Promotions