missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ATP6AP1 (Human) Recombinant Protein
Human ATP6AP1 full-length ORF ( NP_001174.2, 1 a.a. - 470 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000537-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MMAAMATARVRMGPRCAQALWRMPWLPVFLSLAAAAAAAAAEQQVPLVLWSSDRDLWAPAADTHEGHITSDLQLSTYLDPALELGPRNVLLFLQDKLSIEDFTAYGGVFGNKQDSAFSNLENALDLAPSSLVLPAVDWYAVSTLTTYLQEKLGASPLHVDLATLRELKLNASLPALLLIRLPYTASSGLMAPREVLTGNDEVIGQVLSTLKSEDVPYTAALTAVRPSRVARDVAVVAGGLGRQLLQKQPVSPVIHPPVSYNDTAPRILFWAQNFSVAYKDQWEDLTPLTFGVQELNLTGSFWNDSFARLSLTYERLFGTTVTFKFILANRLYPVSARHWFTMERLEVHSNGSVAYFNASQVTGPSIYSFHCEYVSSLSKKGSLLVARTQPSPWQMMLQDFQIQAFNVMGEQFSYASDCASFFSPGIWMGLLTSLFMLFIFTYGLHMILSLKTMDRFDDHKGPTISLTQIV
Specifications
NP_001174.2 | |
ATPase, H+ transporting, lysosomal accessory protein 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATP6AP1 | |
GST |
537 | |
Wheat germ expression system | |
25 μg | |
16A, ATP6IP1, ATP6S1, Ac45, CF2, MGC129781, VATPS1, XAP-3, XAP3 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |