Learn More
ATAGO™ Master-M Refractometer
The MASTER-M series is a good alternative choice when the water resistant feature is not a priority and the Temperature Compensation will be carried out manually. The ATAGO™ Master-M Refractometer is not equipped with the Automatic Temperature Compensation feature.
Brand: ATAGO™ 2313
Additional Details : Weight : 0.80000kg
Description
The MASTER-M is made of metal; for salty and acidic samples, choose the plastic body model (MASTER-PM).
Specifications
Refractometer | |
Master-M | |
Min. Scale ±0.2% |
0.0 to 33.0 Brix | |
± 0.2% | |
Brix |