missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D5599 Revised Oxygenates Mixture, Dr. Ehrenstorfer

Product Code. 30320687
Click to view available options
Quantity:
5 x 2 mL
Unit Size:
10mL
This item is not returnable. View return policy

Product Code. 30320687

Brand: Dr. Ehrenstorfer DREGS09000464

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethanol∣Ethyl tert-Butyl Ether (ETBE)∣Methanol∣Methyl tert-butyl ether∣Regular Unleaded Gasoline∣tert-Amyl Methyl Ether (TAME)
CAS 64-17-5, 637-92-3, 67-56-1, 1634-04-4, 8006-61-9, 994-05-8
Analyte or Component Names Ethanol∣Ethyl tert-Butyl Ether (ETBE)∣Methanol∣Methyl tert-butyl ether∣Regular Unleaded Gasoline∣tert-Amyl Methyl Ether (TAME)
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Quantity 5 x 2 mL
Concentration or Composition (by Analyte or Components) Ethyl tert-Butyl Ether (ETBE) 2 %∣Methanol 2 %∣tert-Amyl Methyl Ether (TAME) 2 %∣Ethanol 10 %∣Methyl tert-butyl ether 14 %∣Regular Unleaded Gasoline 70 %
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.