Learn More
Hirschmann™ Aspirette™ Vacuum-tweezer
Position and pick up objects with ease. Hirschmann™ Aspirette™ Vacuum-tweezer is perfect for picking up and positioning diverse plane objects up to a maximum weight of 70 grams.
Brand:  Hirschmann™ 9901501
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 10088890
  • 235.00 € / Each
Estimated Shipment: 30-11-2021
Add to basket
Description
Description
  • Complete with 1 power supply unit and a charger
  • 3 suction nozzles and suction cups with 3.2 mm, 6 mm and 10mm diameter
  • Battery operated
Specifications
Specifications
9901501
Electric Vacuum Tweezers