missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ASB9 Recombinant Protein

Brand:  Abnova™ H00140462-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138193

  • 508.00 € / 25µg
Estimated Shipment: 21-08-2024
to see stock.

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH01244
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
53.8
8
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ASB9
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
140462
ASB9 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
DKFZp564L0862/FLJ20636/MGC4954
ASB9
Wheat Germ (in vitro)
GST
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions