missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGDIA Recombinant Protein, Abnova
Marke: Abnova H00000396-P01.10ug
Weitere Details : Gewicht : 0.00010kg
Beschreibung
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spezifikation
AAH16031 | |
Solution | |
396 | |
ARHGDIA (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
GDIA1|MGC117248|RHOGDI|RHOGDI-1 | |
ARHGDIA | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
48.18 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARHGDIA | |
Human | |
Yes |