Learn More
Invitrogen™ Centri-Spin™ Spin Columns
Process samples faster than ultrafiltration columns
Brand: Invitrogen™ 4367827
Additional Details : Weight : 0.20410kg
Description
- Centri-Spin™ columns provide excellent recovery of RNA and DNA fragments for a variety of purification applications. Use Centri-Spin columns to remove primers, probes, and dNTPs from DNA, RNA, PCR and end-labeled products
- Rapid and efficient separations
- Permits user to determine buffer composition
- Convenient 20-50μL sample size
- Dry gel format
- Room Temperature Storage
- Microcentrifuge format
- Excellent Recovery
- Centri-Spin columns provide excellent recovery of DNA fragments with sizes greater than 10, 20 or 40 base pairs (depending on kit), while removing >98% of salts, NTP's and other low-molecular-weight impurities. Proteins, peptides, and protein conjugates >5kD can also be efficiently separated/desalted from unwanted low-molecular-weight impurities
- Designed for Performance
- The column design is based on the description of gel filtration for the purification of DNA from nick translation reactions by Sambrook, et.al. Each Centri-Spin consists of a specially fritted microfuge tube, dry gel, a wash tube and a sample collection tube, all specifically designed for this purpose
- Versatile
- The column gel can be hydrated with either reagent-grade water or a suitable buffer, and spun in a microcentrifuge or swinging-bucket centrifuge to remove the intersititial fluid. After the sample is applied, spin the column again to remove the low-molecular-weight components, leaving a purified sample exchanged into the selected buffer
DNA & RNA Purification & Analysis, DNA Extraction, PCR Product Clean-Up
Specifications
Centri-Spin 10 | |
20 Columns |