Learn More
Invitrogen™ Anza™ 51 BspTI
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Brand:  Invitrogen™ IVGN0516
62.33 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.10000kg
Product Code. 15543954
  • 93.50 € / 1500 units
Estimated Shipment: 11-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
For Research Use Only. All usage must comply with product instructions.
Includes: Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer
Description
Description
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
  • One buffer for all restriction enzymes
  • One digestion protocol for all DNA types
  • Complete digestion in 15 min
  • Overnight digestion without star activity
Certifications
CoA
Specifications
Specifications
300 μL Anza 51 BspTI
1 mL Anza Buffer (10X)
1 mL Anza Red Buffer (10X)
Store at -20°C.
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Anza™ 10X Red Buffer, Anza™ 10X Buffer
37°C
Restriction Enzyme Cloning System
Anza™
Approved for shipment on Wet or Dry Ice
BspTI
No
No
Traditional Cloning