missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZP4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539556
Additional Details : Weight : 0.00900kg
Description
The mammalian zona pellucida is composed of four major glycoproteins, ZP1, ZP2, ZP3 and ZP4, which may act as sperm receptors. Two forms of porcine ZP4 peptides exist: one consisting of 128 amino acid residues and the other of 133 amino acid residues. These two peptides are identical, except the larger form contains an additional five amino acid sequence at its carboxy-terminal end. Both peptides have two potential N-linked glycosylation sites. The smaller peptide shares 39.1% identity with the amino-terminal region of mouse ZP2 polypeptide. Based on results from animal studies, ZP4 antigen is a promising candidate for the development of a contraceptive vaccine.Specifications
ZP4 | |
Polyclonal | |
Unconjugated | |
ZP4 | |
Zona pellucida sperm-binding protein 4, Zona pellucida glycoprotein 4, Zp-4, Zona pellucida protein B, Processed zona pellucida sperm-binding protein 4, ZPB | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
282833, 57829 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q12836, Q8CH34 | |
ZP4 | |
A synthetic peptide derived from the internal region of human ZP4 | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |