missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIRAP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA112815
Additional Details : Weight : 0.01000kg
Description
A suggested positive control for this product is liver, kidney, spleen, skeletal muscle or heart lysate.
Toll-like receptors (TLRs) are evolutionarily conserved patternrecognition molecules that recognize different microbial products during infection and serve as an important link between the innate and adaptive immune responses. The signaling of these TLRs is kept under tight control by the expression of endogenous inhibiting proteins such as RP105, a recently identified homolog of TLR4. This protein, in association with MD-1, interacts with and inhibits the TLR4/MD-2 signaling pathway.. While MD-2 can directly bind to LPS, one of the activating molecules of TLR4, the function of MD -1 is less well-known. It has been suggested however, that the RP105/MD-1 complex influences antibody production mediated by both TLR4/MD-2 and TLR2 receptor complexes.Specifications
TIRAP | |
Polyclonal | |
PBS with 0.02% sodium azide | |
P58753, Q99JY1 | |
TIRAP | |
Synthetic peptide corresponding to 15 residues near the C-terminus of mouse TIRAP. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
TIRAP | |
Mal, wyatt | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114609, 117149, 680127 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |