Learn More
TFF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533074
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla (100%); Gibbon, Marmoset, Cat, Horse (90%); Monkey, Mouse, Pig (80%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
TFF2 Polyclonal antibody specifically detects TFF2 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
TFF2 | |
Polyclonal | |
Unconjugated | |
Tff2 | |
mSP; pancreatic spasmolytic polypeptide; PSP; SML1; SP; spasmolysin; Spasmolytic polypeptide; spasmolytic protein 1; Tff2; Trefoil factor 2; trefoil factor 2 (spasmolytic protein 1) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7032 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 0.1% sodium azide | |
Q03403 | |
Tff2 | |
Synthetic 10 amino acid peptide from internal region of human TFF2. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |