Learn More
TCR gamma/delta Monoclonal Antibody (5A6.E9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen TCR1061
Additional Details : Weight : 0.68040kg
Description
TCR1061 targets TCR Pan TCR gamma delta in FACS, IHC(F), IP, and TCA applications and shows reactivity with Human samples. The clone 5A6.E9 has been listed as 5A6.E91 in some publications and documents. These are the same clone. The TCR1061 immunogen is human PEER cells. TCR1061 detects TCR Pan TCR gamma delta which has a predicted molecular weight of approximately 18 kDa.
TCR gamma/delta Monoclonal antibody specifically detects TCR gamma/delta in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, T-Cell Activation.Specifications
TCR gamma/delta | |
Monoclonal | |
0.15 mg/mL | |
PBS with 0.5% BSA and 0.1% sodium azide | |
0 | |
TRD, Trg | |
Human leukemic PEER cells | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, T-Cell Activation | |
5A6.E9 | |
Unconjugated | |
Trg | |
RGD1311300; similar to T cell receptor V delta 6; T cell receptor gamma locus; T3/TCR complex; TCR delta gamma; TCR gamma/delta; TCR-CD3 complex; Tcrg; Trg; Trg@ | |
Mouse | |
Protein A | |
RUO | |
6964, 6965 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |