Learn More
SP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527243
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (100%), Rat (100%), Chicken (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SP1 Polyclonal antibody specifically detects SP1 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
SP1 | |
Polyclonal | |
Unconjugated | |
SP1 | |
1110003E12Rik; AA450830; AI845540; HGNC:11205; SP1; Sp1 transcription factor; Sp1-1; specificity protein 1; trans-acting transcription factor 1; transcription factor Sp1; TSFP1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6667 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P08047 | |
SP1 | |
Synthetic peptide corresponding to a region within amino acids 470 and 564 of Human SP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |