Learn More
SMAD4 Monoclonal Antibody (4G1C6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515682
Additional Details : Weight : 0.01000kg
Description
MA5-15682 targets SMAD4 in indirect ELISA, FACS, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15682 immunogen is purified recombinant fragment of human SMAD4 expressed in E. Coli. . MA5-15682 detects SMAD4 which has a predicted molecular weight of approximately 65kDa.
SMAD4 Monoclonal antibody specifically detects SMAD4 in Human, Mouse samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
SMAD4 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P97471, Q13485 | |
SMAD4 | |
Purified recombinant fragment of human SMAD4 expressed in E. Coli. | |
RUO | |
17128, 4089 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
4G1C6 | |
Unconjugated | |
SMAD4 | |
AW743858; D18Wsu70e; Deleted in Pancreatic Carcinoma 4 (DPC4); deleted in pancreatic carcinoma locus 4; deletion target in pancreatic carcinoma 4; Deletion target in pancreatic carcinoma 4 homolog; DPC4; hSMAD4; JIP; MAD (mothers against decapentaplegic Drosophila) homolog 4; MAD homolog 4; MAD homolog 4 (MADH4); Madh4; mothers against decapentaplegic homolog 4; Mothers against decapentaplegic homolog 4 (MADH4); mothers against decapentaplegic, Drosophila, homolog of, 4; mothers against DPP homolog 4; MYHRS; SMAD; SMAD 4; SMAD family member 4; SMAD, mothers against DPP homolog 4; SMAD4 | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |