missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Sheep Red Blood Cells, Polyclonal, Antiserum, MP Biomedicals
Brand: MP Biomedicals 0855800
153.03 EUR valid until 2024-12-31
Use promo code "23257" to get your promotional price.
Additional Details : Weight : 0.00200kg
Description
Host Species: Rabbit
Species Reactivity: Sheep
Specifications
Red blood cells | |
Unconjugated | |
2 mL | |
Sheep |
Polyclonal | |
Rabbit | |
Primary | |
Lyophilized |