missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCXA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523943
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with chicken based on sequence homology.
SCXA Polyclonal antibody specifically detects SCXA in Human, Mouse samples. It is validated for Western Blot.Specifications
SCXA | |
Polyclonal | |
Unconjugated | |
SCX | |
basic helix-loop-helix transcription factor scleraxis; BB114693; Bhlha41; BHLHA48; class A basic helix-loop-helix protein 41; Class A basic helix-loop-helix protein 48; class II bHLH protein scleraxis; Scl; scleraxis; scleraxis basic helix-loop-helix transcription factor; scleraxis bHLH transcription factor; scleraxis homolog A; scleraxis homolog B; SCX; SCXA; SCXB | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
20289, 642658 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q64124, Q7RTU7 | |
SCX | |
KLH conjugated synthetic peptide between 172-201 amino acids from the C-terminal region of human SCXA | |
200 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |