missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCUBE3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534427
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is mouse kidney tissue lysate. PA5-34427 can be used with blocking peptide PEP-1469.
SCUBE3 Polyclonal antibody specifically detects SCUBE3 in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blot.Specifications
SCUBE3 | |
Polyclonal | |
Unconjugated | |
SCUBE3 | |
CEGF3; CUB domain and EGF-like repeat containing 3; D030038I21Rik; SCUBE3; signal peptide; signal peptide, CUB and EGF-like domain containing protein 3; signal peptide, CUB and EGF-like domain-containing protein 3; signal peptide, CUB domain and EGF like domain containing 3; signal peptide, CUB domain, EGF-like 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
222663, 268935 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q66PY1, Q8IX30 | |
SCUBE3 | |
A 15 amino acid synthetic peptide near the center of human SCUBE3 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |