Learn More
hnRNP M3-M4 Monoclonal Antibody (2A6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA134962
Additional Details : Weight : 0.01000kg
Description
MA1-34962 detects RNP M3-M4 from mouse, rat, human, and rabbit RNP M1-M4 samples. MA1-34962 has been successfully used in immunofluorescence, immunoprecipitation, and Western blot applications. The MA1-34962 immunogen is M19 fusion protein containing full-length human protein. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Specifications
hnRNP M3-M4 | |
Monoclonal | |
Unconjugated | |
P52272, Q9D0E1, Q62826 | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse, Rabbit, Rat | |
Antibody | |
IgG2b κ |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
2A6 | |
HNRNPM | |
HNRNPM | |
M19 fusion protein containing full-length human protein. | |
RUO | |
100356502, 116655, 4670, 76936 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |