missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL5 (RANTES) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517085
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
CCL5 (RANTES) Polyclonal antibody specifically detects CCL5 (RANTES) in Human, Non-human Primate samples. It is validated for Western Blot.Specifications
CCL5 (RANTES) | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P13501 | |
CCL5 | |
Synthetic peptide corresponding to residues surrounding Arg40 of human RANTES | |
100 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CCL5 | |
beta-chemokine RANTES; CC chemokine; C-C motif chemokine 5; C-C motif chemokine ligand 5; CCL5; chemokine (C-C motif) ligand 5; D17S136E; EoCP; eosinophil chemotactic cytokine; MGC17164; MuRantes; OTTHUMP00000197106; RANTES; RANTES protein; RANTES(3-68); RANTES(4-68); regulated upon activation normal T-cell expressed and secreted; regulated upon activation, normally T-expressed, and presumably secreted; Scya5; SIS d; SISd; SIS-delta; SISd; small inducible cytokine A5; small inducible cytokine subfamily A (Cys-Cys), member 5; small-inducible cytokine A5; T cell-specific protein P228; T-cell specific protein p288; T-cell-specific protein RANTES; TCP228; Unknown (protein for MGC:127014) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6352 | |
-20°C | |
Liquid |