Learn More
Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Rhodamine Red™-X, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen R6394
Additional Details : Weight : 0.00454kg
Description
Rabbit IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Rabbit IgG (H+L) Cross-Adsorbed in Rabbit samples. It is validated for Immunocytochemistry.Specifications
Rabbit IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Rhodamine Red-X | |
Goat | |
purified | |
RUO | |
Rabbit | |
Liquid |
Immunocytochemistry | |
2 mg/mL | |
PBS with 5mM sodium azide; pH 7.5 | |
Gamma Immunoglobins Heavy and Light chains | |
1 mg | |
Secondary | |
4° C, store in dark | |
IgG |