missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-c-Kit (Tyr823) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 44498G
Additional Details : Weight : 0.02000kg
Description
Phospho-c-Kit (Tyr823) Polyclonal antibody specifically detects Phospho-c-Kit (Tyr823) in Avian, Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Phospho-c-Kit (Tyr823) | |
Polyclonal | |
Dulbecco's PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide; pH 7.3 | |
P10721 | |
KIT | |
The antiserum was produced against a chemically synthesized phosphopeptide derived from a region of human c-Kit that contains tyrosine 823. The sequence is conserved in cow and dog. | |
100 μL | |
Primary | |
Avian, Human | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
Unconjugated | |
KIT | |
belly-spot; Bs; CD117; ckit; C-Kit; c-KIT gene1; c-kit protein; c-kit proto-oncogene protein; c-kit receptor; c-kit receptor tyrosine kinase; dominant spotting; Dominant white spotting; Fdc; Gsfsco1; Gsfsco5; Gsfsow3; KIT; kit oncogene; KIT proto-oncogene receptor tyrosine kinase; KIT proto-oncogene, receptor tyrosine kinase; mast cell growth factor receptor; mast/stem cell growth factor receptor; mast/stem cell growth factor receptor Kit; MGF; p145 c-kit; PBT; Piebald trait protein; protein kinase; proto-oncogene c-Kit; proto-oncogene tyrosine-protein kinase Kit; receptor tyrosine kinase c-kit; receptor tyrosine kinase type III; SCFR; SCO1; SCO5; Sl; soluble KIT variant 1; SOW3; spotted sterile male; Ssm; Steel Factor Receptor; Tr-kit; tyrosine kinase receptor; tyrosine kinase receptor c-KIT; tyrosine-protein kinase Kit; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein; W | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3815 | |
-20°C | |
Liquid |